Tryptamine, 98+%, Thermo Scientific Chemicals
Tryptamine, 98+%, Thermo Scientific Chemicals
Tryptamine, 98+%, Thermo Scientific Chemicals
Tryptamine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tryptamine, 98+%, Thermo Scientific Chemicals
CAS: 61-54-1 | C10H12N2 | 160.22 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11116.0910 g
Catalog number ALFA11116.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS1530-03-6
IUPAC Name(1-phenylpropyl)benzene
Molecular FormulaC15H16
InChI KeyBUZMJVBOGDBMGI-UHFFFAOYSA-N
SMILESCCC(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormViscous liquid
GC/MS AnalysisConforms
Appearance (Color)Colorless
Assay (GC)≥98.5%
Refractive Index1.564
Tryptamine is commonly used in the preparation of biologically active compounds such as neurotransmitters and psychedelics, multi-layered analysis, used to differentiate and identify biogenic amines1
  • in the synthesis of indole ring-terminated thiol, ω-mercaptooctyltryptamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tryptamine is commonly used in the preparation of biologically active compounds such as neurotransmitters and psychedelics, multi-layered analysis, used to differentiate and identify biogenic amines1 • in the synthesis of indole ring-terminated thiol, ω-mercaptooctyltryptamide.
Solubility
Soluble in DMSO, Methanol. Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only
General References:
  1. J. H. GADDUM,; Z. P. PICARELLI.TWO KINDS OF TRYPTAMINE RECEPTOR . British Journal of Pharmacology. 1997, 120 (S1),134-139.
  2. Gaddum J. H. Tryptamine receptors. The Journal of Physiology. 1953, 119(2-3),363-368.