Antipyrine, 98%, Thermo Scientific Chemicals
Antipyrine, 98%, Thermo Scientific Chemicals
Antipyrine, 98%, Thermo Scientific Chemicals
Antipyrine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Antipyrine, 98%, Thermo Scientific Chemicals
CAS: 60-80-0 | C11H12N2O | 188.23 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11089.36500 g
Catalog number ALFA11089.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS589-16-2
IUPAC Name4-ethylaniline
Molecular FormulaC8H11N
InChI KeyHRXZRAXKKNUKRF-UHFFFAOYSA-N
SMILESCCC1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Form)Liquid
Appearance (Color)Clear yellow to red-brown
Infrared spectrumConforms
Refractive index1.5532 to 1.5552 (20°C, 589 nm)