DL-Lysine monohydrochloride, 99%
DL-Lysine monohydrochloride, 99%
DL-Lysine monohydrochloride, 99%
DL-Lysine monohydrochloride, 99%
Thermo Scientific Chemicals
DL-Lysine monohydrochloride, 99%
CAS: 70-53-1 | C6H15ClN2O2 | 182.65 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11066.22100 g
Catalog number ALFA11066.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS10424-65-4
IUPAC Nametetramethylazanium pentahydrate hydroxide
Molecular FormulaC4H23NO6
InChI KeyMYXKPFMQWULLOH-UHFFFAOYSA-M
SMILESO.O.O.O.O.[OH-].C[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Hygroscopic powder or crystals
Melting point62°C to 73°C
Titration with HCl>=96.0 %
Appearance (Color)White to off-white
DL-Lysine monohydrochloride acts as an intermediate in the preparation of 3-amino-azepan-2-one. It also serves as an essential amino acid. It is used as animal feed. In addition to this, it is used in the construction of polylysine polymers, which finds application in lysine-based polypeptide gels and films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DL-Lysine monohydrochloride acts as an intermediate in the preparation of 3-amino-azepan-2-one. It also serves as an essential amino acid. It is used as animal feed. In addition to this, it is used in the construction of polylysine polymers, which finds application in lysine-based polypeptide gels and films.
Solubility
Soluble in water. Insoluble in ethanol, ethyl ether, acetone, benzene and common neutral solvents.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Serrano, N.; González-Calabuig, A.; del Valle, M. Crown ether-modified electrodes for the simultaneous stripping voltammetric determination of Cd(II), Pb(II) and Cu(II). Talanta 2015, 138, 130-137.
  2. Metkar, P. S.; Scialdone, M. A.; Moloy, K. G. Lysinol: a renewably resourced alternative to petrochemical polyamines and aminoalcohols. Green Chem. 2014, 16 (10), 4575-4586.