Tri-n-octylphosphine oxide, 98%, Thermo Scientific Chemicals
Tri-n-octylphosphine oxide, 98%, Thermo Scientific Chemicals
Tri-n-octylphosphine oxide, 98%, Thermo Scientific Chemicals
Tri-n-octylphosphine oxide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tri-n-octylphosphine oxide, 98%, Thermo Scientific Chemicals
CAS: 78-50-2 | C24H51OP | 386.645 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11062.22100 g
Catalog number ALFA11062.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS7758-89-6
IUPAC Nameλ¹-copper(1+) chloride
Molecular FormulaClCu
InChI KeyOXBLHERUFWYNTN-UHFFFAOYSA-M
SMILES[Cl-].[Cu+]
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfate (SO4)=<500 ppm
Appearance (Color)Light green to grey
Titration Complexometric98.5 to 101.5 %
Appearance (Form)Crystalline powder
Iron (Fe)=<200 ppm
Tri-n-octylphosphine oxide is a solvent used in the extraction of metals, especially uranium, zirconium and hafnium. It is also used as an auxiliary reagent in the solvent extraction of metals. It is also used to extract hydrogen bonding organic compounds. Further, it is useful as a capping ligand for the production of quantum dots, such as those consisting of cadmium selenide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tri-n-octylphosphine oxide is a solvent used in the extraction of metals, especially uranium, zirconium and hafnium. It is also used as an auxiliary reagent in the solvent extraction of metals. It is also used to extract hydrogen bonding organic compounds. Further, it is useful as a capping ligand for the production of quantum dots, such as those consisting of cadmium selenide.
Solubility
Soluble in chloroform, toluene and hexane. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Used in the extraction of various metals into organic solvents.
  2. Ren, C.; Hao, J.; Chen, H.; Wang, K.; Wu, D. Prepare core-multishell CdSe/ZnS nanocrystals with pure color and controlled emission by tri-n-octylphosphine-assisted method. Appl. Surf. Sci. 2015, 353, 480-488.
  3. Grimes, T. S.; Zalupski, P. R.; Martin, L. R. Features of the Thermodynamics of Trivalent Lanthanide/Actinide Distribution Reactions by Tri-n-octylphosphine Oxide and Bis(2-ethylhexyl) Phosphoric Acid. J. Phys. Chem. B 2014, 118 (44), 12725-12733.