Tryptamine hydrochloride, 98+%, Thermo Scientific Chemicals
Tryptamine hydrochloride, 98+%, Thermo Scientific Chemicals
Tryptamine hydrochloride, 98+%, Thermo Scientific Chemicals
Tryptamine hydrochloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tryptamine hydrochloride, 98+%, Thermo Scientific Chemicals
CAS: 343-94-2 | C10H13ClN2 | 196.678 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11030.1850 g
Catalog number ALFA11030.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS17524-05-9
IUPAC Namedioxomolybdenumbis(ylium) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14MoO6
InChI KeySKMUJBBRXZPAJY-VGKOASNMSA-L
SMILESO=[Mo++]=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to grey-green to light brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Elemental Analysis>=96.0 %