3-Iodotoluene, 99%, Thermo Scientific Chemicals
3-Iodotoluene, 99%, Thermo Scientific Chemicals
3-Iodotoluene, 99%, Thermo Scientific Chemicals
3-Iodotoluene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Iodotoluene, 99%, Thermo Scientific Chemicals
CAS: 625-95-6 | C7H7I | 218.037 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10999.1850 g
Catalog number ALFA10999.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Iodotoluene
CAS625-95-6
Health Hazard 1H227
Health Hazard 2GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P210-P280-P370+P378q-P501c
View more
3-Iodotoluene was used in the preparartion of indene derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Iodotoluene was used in the preparartion of indene derivatives.
Solubility
Not miscible in water. Soluble in benzene, alcohol and ether.
Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Zheng-Hui Guan; Zhi-Hui Ren; Lian-Biao Zhao; Yong-Min Liang. Palladium-catalyzed synthesis of indene derivatives via propargylic carbonates with in situ generated organozinc compounds. Organic & Biomolecular Chemistry. 2008, 6,(6), 1040-1045.
  2. O.Buchman; G.Y.Cao; C.T.Peng. Structure assignment by retention index in gas-liquid radiochromatography of substituted cyclohexenes. Journal of Chromatography A. 1984, 312,75-90.