Fumaric acid, 99%, Thermo Scientific Chemicals
Fumaric acid, 99%, Thermo Scientific Chemicals
Fumaric acid, 99%, Thermo Scientific Chemicals
Fumaric acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Fumaric acid, 99%, Thermo Scientific Chemicals
CAS: 110-17-8 | C4H4O4 | 116.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
500 g
10,000 g
Catalog number ALFA10976.0E
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialFumaric acid
CAS110-17-8
Health Hazard 1H319-H500
Health Hazard 2GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3P264b-P280i-P305+P351+P338
View more
Fumaric acid is used as a food additive and acid regulator. It is used in the production of polyester resins and polyhydric alcohols. It is also used as a mordant for dyes, paint additives, coating additives, plasticizers, processing aids and specific to petroleum production. Further, it is used in dentifrices as a stain remover and to make other chemicals. In addition to this, it is used in rosin esters, drying oils, printing inks and flavoring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Fumaric acid is used as a food additive and acid regulator. It is used in the production of polyester resins and polyhydric alcohols. It is also used as a mordant for dyes, paint additives, coating additives, plasticizers, processing aids and specific to petroleum production. Further, it is used in dentifrices as a stain remover and to make other chemicals. In addition to this, it is used in rosin esters, drying oils, printing inks and flavoring agent.
Solubility
Soluble in water, ethanol and concentrated sulfuric acid. Slightly soluble in ethyl ether and acetone. Insoluble in chloroform, olive oil, carbon tetrachloride, xylene, molten camphor, liquid ammonia and benzene.
Notes
Incompatible with oxidizing agents, amines and strong bases.
RUO – Research Use Only
General References:
  1. Rose, J. L.; Tata, B. V. R.; Talmon, Y.; Aswal, V. K.; Hassan, P. A.; Sreejith, L. Distinctive effect of maleic acid and fumaric acid on structural transitions in cationic micellar solution. Colloid. Polym. Sci. 2015, 293 (5), 1383-1390.
  2. Huang, D.; Wang, R.; Du, W.; Wang, G.; Xia, M. Activation of glycerol metabolic pathway by evolutionary engineering of Rhizopus oryzae to strengthen the fumaric acid biosynthesis from crude glycerol. Bioresour. Technol. 2015, 196, 263-272.