Furfuryl alcohol, 98%, Thermo Scientific Chemicals
Furfuryl alcohol, 98%, Thermo Scientific Chemicals
Furfuryl alcohol, 98%, Thermo Scientific Chemicals
Furfuryl alcohol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Furfuryl alcohol, 98%, Thermo Scientific Chemicals
CAS: 98-00-0 | C5H6O2 | 98.101 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALFA10968.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS17522-69-9
IUPAC Nameneodymium(3+) triperchlorate
Molecular FormulaCl3NdO12
InChI KeyHBKMAYJLXKBOER-UHFFFAOYSA-K
SMILES[Nd+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Form50% Aqueous Solution
Assay from Suppliers CofANeodymium content: 12.6-13.6%
Chloride content≤0.01%
Insoluble material≤0.01%
Furfuryl alcohol was used in the synthesis of continuous titanium carbide nanofibers/nanoribbon. Furfuryl alcohol is an efficient trapping agent for singlet oxygen determination in natural waters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Furfuryl alcohol was used in the synthesis of continuous titanium carbide nanofibers/nanoribbon. Furfuryl alcohol is an efficient trapping agent for singlet oxygen determination in natural waters.
Solubility
Soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep containers tightly closed in a cool, well-ventilated place. Keep in properly labeled containers. Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Werner R. Haag.; Jürg Hoigne.; Ernst Gassman.; André M. Braun. Singlet oxygen in surface waters — Part I: Furfuryl alcohol as a trapping agent. Chemosphere. 1984, 13, (5-6), 631-640.
  2. Mekki Choura.; Naceur M. Belgacem.; Alessandro Gandini. Acid-Catalyzed Polycondensation of Furfuryl Alcohol:  Mechanisms of Chromophore Formation and Cross-Linking. Macromolecules. 1996, 29, (11), 3839-3850.