Diethylenetriaminepentaacetic acid, 98+%, Thermo Scientific Chemicals
Diethylenetriaminepentaacetic acid, 98+%, Thermo Scientific Chemicals
Diethylenetriaminepentaacetic acid, 98+%, Thermo Scientific Chemicals
Diethylenetriaminepentaacetic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethylenetriaminepentaacetic acid, 98+%, Thermo Scientific Chemicals
CAS: 67-43-6 | C14H23N3O10 | 393.349 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFA10926.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDiethylenetriaminepentaacetic acid
CAS67-43-6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Diethylenetriaminepentaacetic acid is used as chelating agent and forms complexes with metal ions. It finds application as a magnetic resonance imaging (MRI) contrasting agent, since it improves the image by forming a complex with a gadolinium ion. It can also be used for treatment of radioactive materials such as plutonium, americium and other actinides. It is an active component of manganese and zinc fertilizers used in agrochemical industry. It acts as a color inhibitor and is used in the production of acrylon. It is also used in soaps as a water softener.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethylenetriaminepentaacetic acid is used as chelating agent and forms complexes with metal ions. It finds application as a magnetic resonance imaging (MRI) contrasting agent, since it improves the image by forming a complex with a gadolinium ion. It can also be used for treatment of radioactive materials such as plutonium, americium and other actinides. It is an active component of manganese and zinc fertilizers used in agrochemical industry. It acts as a color inhibitor and is used in the production of acrylon. It is also used in soaps as a water softener.
Solubility
Soluble in water and alkali.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Chelating agent.
  2. Fitzsimmons, J.; Jay, M. Preparative-scale separation of ethyl esters of diethylenetriaminepentaacetic acid by flash chromatography. J. Radioanal. Nucl. Chem. 2015, 305 (1), 329-336.
  3. Bhatt, R.; Sreedhar, B.; Padmaja, P. Adsorption of chromium from aqueous solutions using crosslinked chitosan-diethylenetriaminepentaacetic acid. Int. J. Biol. Macromol. 2015, 74, 458-466.