Glycerol tripalmitate, 98%, Thermo Scientific Chemicals
Glycerol tripalmitate, 98%, Thermo Scientific Chemicals
Glycerol tripalmitate, 98%, Thermo Scientific Chemicals
Glycerol tripalmitate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycerol tripalmitate, 98%, Thermo Scientific Chemicals
CAS: 555-44-2 | C51H98O6 | 807.339 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA10922.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGlycerol tripalmitate
CAS555-44-2
Melting Point60°C to 64°C
Recommended StorageStore at -20°C
RTECS NumberRT4953500