Glycerol tripalmitate, 98%, Thermo Scientific Chemicals
Glycerol tripalmitate, 98%, Thermo Scientific Chemicals
Glycerol tripalmitate, 98%, Thermo Scientific Chemicals
Glycerol tripalmitate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycerol tripalmitate, 98%, Thermo Scientific Chemicals
CAS: 555-44-2 | C51H98O6 | 807.339 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFA10922.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight807.34