2,3-Dimethylaniline, 98%, Thermo Scientific Chemicals
2,3-Dimethylaniline, 98%, Thermo Scientific Chemicals
2,3-Dimethylaniline, 98%, Thermo Scientific Chemicals
2,3-Dimethylaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,3-Dimethylaniline, 98%, Thermo Scientific Chemicals
CAS: 87-59-2 | C8H11N | 121.183 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
Catalog number ALFA10918.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS7748-36-9
IUPAC Nameoxetan-3-ol
Molecular FormulaC3H6O2
InChI KeyQMLWSAXEQSBAAQ-UHFFFAOYSA-N
SMILESOC1COC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=94.0 %
Appearance (Color)Clear colorless to light yellow