2,3-Dimethylaniline, 98%, Thermo Scientific Chemicals
2,3-Dimethylaniline, 98%, Thermo Scientific Chemicals
2,3-Dimethylaniline, 98%, Thermo Scientific Chemicals
2,3-Dimethylaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,3-Dimethylaniline, 98%, Thermo Scientific Chemicals
CAS: 87-59-2 | C8H11N | 121.183 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
500 g
Catalog number ALFA10918.0E
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2500 g
Request bulk or custom format
Chemical Identifiers
CAS608-31-1
IUPAC Name2,6-dichloroaniline
Molecular FormulaC6H5Cl2N
InChI KeyJDMFXJULNGEPOI-UHFFFAOYSA-N
SMILESNC1=C(Cl)C=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Melting point35°C to 39°C
GC>=97.5 %
Appearance (Form)Crystalline solid
AppearanceMay darken on storage