Sodium tetraphenylborate, 99%, Thermo Scientific Chemicals
Sodium tetraphenylborate, 99%, Thermo Scientific Chemicals
Sodium tetraphenylborate, 99%, Thermo Scientific Chemicals
Sodium tetraphenylborate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium tetraphenylborate, 99%, Thermo Scientific Chemicals
CAS: 143-66-8 | C24H20BNa | 342.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA10909.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS141-79-7
IUPAC Name4-methylpent-3-en-2-one
Molecular FormulaC6H10O
InChI KeySHOJXDKTYKFBRD-UHFFFAOYSA-N
SMILESCC(C)=CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥90.0% (major component)
Identification (FTIR)Conforms
Refractive Index1.4415-1.4465 @ 20?C
Sodium tetraphenylborate is used as a precipitating reagent for potassium and as a protecting agent to protect amino acids. It is used in the preparation of N-acylammonium salts. It finds application as a phenyl donor in palladium-catalyzed cross-coupling reactions to prepare arylalkenes and biaryl compounds. It is used in inorganic and organometallic chemistry due to its solubility in nonpolar solvents. Furthermore, it is involved in the isolation of complexes having dinitrogen ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium tetraphenylborate is used as a precipitating reagent for potassium and as a protecting agent to protect amino acids. It is used in the preparation of N-acylammonium salts. It finds application as a phenyl donor in palladium-catalyzed cross-coupling reactions to prepare arylalkenes and biaryl compounds. It is used in inorganic and organometallic chemistry due to its solubility in nonpolar solvents. Furthermore, it is involved in the isolation of complexes having dinitrogen ligands.
Solubility
Soluble in water and ethanol.
Notes
Light sensitive. Incompatible with strong acids and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Reagent for potassium by precipitation: Anal. Chem., 29, 1044 (1957).
  2. Has been used to protect amino acids by formation of a diphenylborinate chelate, stable to AcOH or TFA, but cleaved by base: Tetrahedron, 39, 2995 (1983); Liebigs Ann. Chem., 127 (1989):
  3. The cross coupling of tetraphenylborates or aryl boronic acids with aryl halides is catalyzed by Pd(OAc)2 in aqueous solutions: Dokl. Chem. (Engl. Transl.), 315, 354 (1990); see also: Gazz. Chem. Ital., 120, 779 (1990). Cross couples with vinyl and aryl triflates under Pd catalysis: Tetrahedron Lett., 35, 4835 (1992); 33, 4815 (1992). Similarly, allyl acetates are phenylated: Tetrahedron Lett., 31, 7453 (1990).
  4. Howa, J. D.; Lott, M. J.; Ehleringer, J. R. Isolation and stable nitrogen isotope analysis of ammonium ions in ammonium nitrate prills using sodium tetraphenylborate. Rapid Commun. Mass Spectrom. 2014, 28 (13), 1530-1534.
  5. Boruń, A.; Trzcińska, I.; Bald, A. Conductometric Studies of Sodium Iodide, Sodium Tetraphenylborate, Tetrabutylammonium Iodide, and Sodium Tetrafluoroborate in 1-Propanol at Temperatures from (283.15 to 318.15) K. Int. J. Electrochem. Sci. 2014, 9, 7805-7818.