Aluminum potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals
Aluminum potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals
Aluminum potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals
Aluminum potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aluminum potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals
CAS: 7784-24-9 | AlH24KO20S2 | 474.372 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
Catalog number ALFA10906.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAluminum potassium sulfate dodecahydrate
Melting Point92°C
CAS7784-24-9
Recommended StorageAmbient temperatures
RTECS NumberWS5690000