Antimony potassium tartrate hydrate, 98%, Thermo Scientific Chemicals
Antimony potassium tartrate hydrate, 98%, Thermo Scientific Chemicals
Antimony potassium tartrate hydrate, 98%, Thermo Scientific Chemicals
Antimony potassium tartrate hydrate, 98%, Thermo Scientific Chemicals
Antimony potassium tartrate hydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Antimony potassium tartrate hydrate, 98%, Thermo Scientific Chemicals
CAS: 331753-56-1 | C8H4K2O12Sb2 | 613.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFA10889.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS1761-71-3
IUPAC Name4-[(4-aminocyclohexyl)methyl]cyclohexan-1-amine
Molecular FormulaC13H26N2
InChI KeyDZIHTWJGPDVSGE-UHFFFAOYSA-N
SMILESNC1CCC(CC2CCC(N)CC2)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0% (sum of stereoisomers)
FormFused solid or clear liquid as melt
Appearance (Color)White or colorless to pale brown
Identification (FTIR)Conforms
Antimony potassium tartrate hydrate is a compound used in the treatment of schistosomiasis. It is also used in the textile industry to aid in binding certain dyes to fabrics. It finds application in medicine as an expectorant and a nauseant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Antimony potassium tartrate hydrate is a compound used in the treatment of schistosomiasis. It is also used in the textile industry to aid in binding certain dyes to fabrics. It finds application in medicine as an expectorant and a nauseant.
Solubility
Soluble in water. Insoluble in ethanol.
Notes
Incompatible with mineral acids, strong bases, carbonates, lead, silver salts and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Vagvala, T. C.; Pandey, S. S.; Ogomi, Y.; Ma, T.; Hayase, S. Investigation of metal xanthates as latent curing catalysts for epoxy resin via formation of in-situ metal sulfides. Inorg. Chim. Acta 2015, 435, 292-298.
  2. Nguyen, T. H.; Müller, R. H.; Taupitz, M.; Schnorr, J.; Hamm, B.; Wagner, S. Novel oral phosphate binder with nanocrystalline maghemite-phosphate binding capacity and pH effect. Int. J. Pharm. 2015, 482 (1), 21-26.