Tetra-n-butylphosphonium bromide, 99%, Thermo Scientific Chemicals
Tetra-n-butylphosphonium bromide, 99%, Thermo Scientific Chemicals
Tetra-n-butylphosphonium bromide, 99%, Thermo Scientific Chemicals
Tetra-n-butylphosphonium bromide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetra-n-butylphosphonium bromide, 99%, Thermo Scientific Chemicals
CAS: 3115-68-2 | C16H36BrP | 339.342 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number ALFA10868.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS993-10-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, pale yellow
FormLiquid
Assay (unspecified)>96%
Tetra-n-butylphosphonium bromide is used as a phase-transfer catalyst at high temperatures, at which nitrogen analogues tend to undergo elimination e.g. as catalyst for the dealkoxycarbonylation of malonates and β-keto esters by heating in stearic acid. used as an ionic liquid solvent in the regioselective O-alkylation of C/O ambident nucleophiles. It is a synthetic reagent used in proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetra-n-butylphosphonium bromide is used as a phase-transfer catalyst at high temperatures, at which nitrogen analogues tend to undergo elimination e.g. as catalyst for the dealkoxycarbonylation of malonates and ß-keto esters by heating in stearic acid. used as an ionic liquid solvent in the regioselective O-alkylation of C/O ambident nucleophiles. It is a synthetic reagent used in proteomics research.
Solubility
Soluble in water (70 g/100 ml)
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature.
RUO – Research Use Only
General References:
  1. Pascal Claina.; Anthony Delahayea.; Laurence Fournaisona.; Nadia Mayoufib.; Didier Dalmazzoneb.; Walter Fürstb. Rheological properties of tetra-n-butylphosphonium bromide hydrate slurry flow. Tetrahedron Letters. 1992, 193-194, 112-122.
  2. Rena M. Pomaville.; Salwa K. Poole.; LaDonna J. Davis.; Colin F. Poole. Solute—solvent interactions in tetra-n-butylphosphonium salts studied by gas chromatography. Chemical Engineering Journal. 1988, 438, 1-14.
  3. Quaternary phosphonium salt, useful as a phase-transfer catalyst at high temperatures, at which nitrogen analogues tend to undergo elimination (see Appendix 2), e.g. as catalyst for the dealkoxycarbonylation of malonates and ß-keto esters by heating in stearic acid: Synthesis, 320 (1985).
  4. Has been used as an ionic liquid solvent in the regioselective O-alkylation of C/O ambident nucleophiles: Tetrahedron Lett., 33, 4435 (1992); see Tetra-n-butyl ammonium bromide, A10249.