2-(Phenylthio)ethanol, 99%, Thermo Scientific Chemicals
2-(Phenylthio)ethanol, 99%, Thermo Scientific Chemicals
2-(Phenylthio)ethanol, 99%, Thermo Scientific Chemicals
2-(Phenylthio)ethanol, 99%, Thermo Scientific Chemicals
2-(Phenylthio)ethanol, 99%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA10861.1850 g
Catalog number ALFA10861.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material2- (Phenylthio)ethanol
CAS699-12-7
Health Hazard 1H302-H312-H315-H319-H332
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c