1-Bromopentane, 99%, Thermo Scientific Chemicals
1-Bromopentane, 99%, Thermo Scientific Chemicals
1-Bromopentane, 99%, Thermo Scientific Chemicals
1-Bromopentane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromopentane, 99%, Thermo Scientific Chemicals
CAS: 110-53-2 | C5H11Br | 151.047 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFA10821.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS69861-71-8
IUPAC Namebis(tris(2-methylphenyl)phosphane) palladium
Molecular FormulaC42H42P2Pd
InChI KeyCUBIJGNGGJBNOC-UHFFFAOYSA-N
SMILES[Pd].CC1=CC=CC=C1P(C1=CC=CC=C1C)C1=CC=CC=C1C.CC1=CC=CC=C1P(C1=CC=CC=C1C)C1=CC=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)C 69.50-71.50%
1-Bromopentane is used in a multitude of organic synthesis. It is used in iron-catalyzed alkylations of aromatic Grignard reagents via cross coupling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromopentane is used in a multitude of organic synthesis. It is used in iron-catalyzed alkylations of aromatic Grignard reagents via cross coupling.
Solubility
Insoluble in water and soluble in alcohol. Miscible in diethyl ether.
Notes
Store away from oxidizing agents and bases.
RUO – Research Use Only
General References:
  1. Keiichi Ohno.; Hiroshi Yoshida.; Hiroatsu Matsuura. Conformational dependence of the isolated C-D stretching and CHD rocking vibrations of selectively monodeuterated alkyl chains as proved by conformational polymorphism of 1-bromopentane. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 1996, 52 (11), 1377-1389.
  2. John P. Idouxa.; Ronald Wysockia.; Sherre Younga.; Jeffrey Turcota.; Charlene Ohlmana.; Russell Leonard. Polymer-Supported Multi-Site Phase Transfer Catalysts. Synthetic Communications: An International Journal for Rapid Communication of Synthetic Organic Chemistry. 1983, 13 (2), 139-144.