4-Phenylphenol, 97%, Thermo Scientific Chemicals
4-Phenylphenol, 97%, Thermo Scientific Chemicals
4-Phenylphenol, 97%, Thermo Scientific Chemicals
4-Phenylphenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Phenylphenol, 97%, Thermo Scientific Chemicals
CAS: 92-69-3 | C12H10O | 170.211 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10817.22100 g
Catalog number ALFA10817.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Phenylphenol
CAS92-69-3
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Fluorescence and phosphorescence quantum yields and fluorescence and phosphorescence lifetimes were obtained for 4-phenylphenol adsorbed on filter paper with either NaCl, NaBr, or NaI at 296 and 93 K. The solid-surface fluorescence and phosphorescence quantum yield values and phosphorescence lifetime values were obtained for p-aminobenzoic acid (PABA) and 4-phenylphenol adsorbed on α-cyclodextrin/NaCl mixtures. 4-Phenylphenol is used as an antioxidant and is a potential EDC.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Fluorescence and phosphorescence quantum yields and fluorescence and phosphorescence lifetimes were obtained for 4-phenylphenol adsorbed on filter paper with either NaCl, NaBr, or NaI at 296 and 93 K. The solid-surface fluorescence and phosphorescence quantum yield values and phosphorescence lifetime values were obtained for p-aminobenzoic acid (PABA) and 4-phenylphenol adsorbed on α-cyclodextrin/NaCl mixtures. 4-Phenylphenol is used as an antioxidant and is a potential EDC.
Solubility
Soluble in water (0.7 mg/ml at 20°C), ether, most common organic solvents, methanol, and chloroform.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Peng Xu,; Jayant Kumar,; Lynne Samuelsonm,; Ashok L. Cholli. Monitoring the Enzymatic Polymerization of 4-Phenylphenol by Matrix-Assisted Laser Desorption Ionization Time-of-Flight Mass Spectrometry: A Novel Approach. Biomacromolecules. 2002, 3(5), 889-893.
  2. S.M. Ramasamy,; R.J. Hurtubise.Temperature effects on the solid-matrix luminescence properties of 4-phenylphenol adsorbed on filter paper. Talanta. 1989, 36(1-2),315-320.