DL-Norleucine, 98%, Thermo Scientific Chemicals
DL-Norleucine, 98%, Thermo Scientific Chemicals
DL-Norleucine, 98%, Thermo Scientific Chemicals
DL-Norleucine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Norleucine, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA10791.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDL-Norleucine
Melting Point>300°C
CAS616-06-8
Recommended StorageAmbient temperatures
EINECS Number210-462-7