(1-Dodecyl)trimethylammonium bromide, 99%, Thermo Scientific Chemicals
(1-Dodecyl)trimethylammonium bromide, 99%, Thermo Scientific Chemicals
(1-Dodecyl)trimethylammonium bromide, 99%, Thermo Scientific Chemicals
(1-Dodecyl)trimethylammonium bromide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(1-Dodecyl)trimethylammonium bromide, 99%, Thermo Scientific Chemicals
CAS: 1119-94-4 | C15H34BrN | 308.348 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10761.1850 g
Catalog number ALFA10761.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS15546-43-7
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
(1-Dodecyl)trimethylammonium bromide is used as a surfactant, as an emulsifier of rubber and asphalt and as an anti-static agent of synthetic fiber. It acts as an ionic surfactant and useful in the preparation of gold nanopartilces along with sodium dodecylsulfate. It is also used as paint strippers and foaming stabilizers. It is used to prepare dodecyltrimethylammonium bromide. By using conductimetry, density measurements, and small angle neutron scattering, it is used to study the micelle formation in water-dimethylsulfoxide mixture. Further, it is a useful biochemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(1-Dodecyl)trimethylammonium bromide is used as a surfactant, as an emulsifier of rubber and asphalt and as an anti-static agent of synthetic fiber. It acts as an ionic surfactant and useful in the preparation of gold nanopartilces along with sodium dodecylsulfate. It is also used as paint strippers and foaming stabilizers. It is used to prepare dodecyltrimethylammonium bromide. By using conductimetry, density measurements, and small angle neutron scattering, it is used to study the micelle formation in water-dimethylsulfoxide mixture. Further, it is a useful biochemical for proteomics research.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with bases and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Wang, P. Y.; Shah, I.; Huang, C. P. Preparation and characterization of functionalized poly(vinyl chloride) membranes for selective separation of perchlorate from water. J. Membr. Sci. 2015, 476, 561-570.
  2. Al-rashdi, A. A. A submicron mesoporous silica for the determination of organosulphur in sea water. Anal. Chem. Res. 2015, 4, 25-32.