Methyl hexanoate, 99%, Thermo Scientific Chemicals
Methyl hexanoate, 99%, Thermo Scientific Chemicals
Methyl hexanoate, 99%, Thermo Scientific Chemicals
Methyl hexanoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl hexanoate, 99%, Thermo Scientific Chemicals
CAS: 106-70-7 | C7H14O2 | 130.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1000 g
Catalog number ALFA10757.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS119-32-4
IUPAC Name4-methyl-3-nitroaniline
Molecular FormulaC7H8N2O2
InChI KeyGDIIPKWHAQGCJF-UHFFFAOYSA-N
SMILESCC1=CC=C(N)C=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Ochre to orange to brown
Infrared spectrumConforms
Appearance (Form)Powder
HPLC>=96.0 %
Melting point74°C to 78°C