(3-Aminopropyl)triethoxysilane, 98%, Thermo Scientific Chemicals
(3-Aminopropyl)triethoxysilane, 98%, Thermo Scientific Chemicals
(3-Aminopropyl)triethoxysilane, 98%, Thermo Scientific Chemicals
(3-Aminopropyl)triethoxysilane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(3-Aminopropyl)triethoxysilane, 98%, Thermo Scientific Chemicals
Alfa Aesar (3-Aminopropyl)triethoxysilane, 98%, Quantity: 100g, Boiling Point: 217 deg.C, CAS: 919-30-2, Molecular Formula: C9H23NO3Si, Molecular Weight: 221.372, Percent Purity: 98%, pH: 11
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number ALFA10668.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight221.37
Unit Size
(3-Aminopropyl)triethoxysilane is useful in the formation of aminopropyl derivative of glass. It acts as an adsorbent for affinity chromatography. It is used to prepare positively charged slides suitable for use with various immunohistochemical and in situ hybridization procedures. In addition, it is used as adhesives and sealant chemicals, paint additives and coating additives. Further, it is used as a silylation reagent for coating glass and silica surfaces and to crosslink and immobilize proteins and other molecules.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(3-Aminopropyl)triethoxysilane is useful in the formation of aminopropyl derivative of glass. It acts as an adsorbent for affinity chromatography. It is used to prepare positively charged slides suitable for use with various immunohistochemical and in situ hybridization procedures. In addition, it is used as adhesives and sealant chemicals, paint additives and coating additives. Further, it is used as a silylation reagent for coating glass and silica surfaces and to crosslink and immobilize proteins and other molecules.
Solubility
Miscible with toluene, acetone, chloroform and ethanol.
Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, acids and moisture.
RUO – Research Use Only
General References:
  1. Huang, J.; Ding, S.; Xiao, W.; Peng, Y.; Deng, S.; Zhang, N. 3-Aminopropyl-triethoxysilane Functionalized Graphene Oxide: A Highly Efficient and Recyclable Catalyst for Knoevenagel Condensation. Catal. Lett. 2015, 145 (4), 1000-1007.
  2. Zhang, N.; Wang, B.; Zhao, C.; Zhang, Y.; Bu, F.; Cui, Y.; Li, X.; Na, H. Dual cross-linked organic-inorganic hybrid polymer electrolyte membranes based on quaternized poly(ether ether ketone) and (3-aminopropyl)triethoxysilane. J. Power Sources 2015, 275, 815-822.