D-Phenylalanine, 99%
D-Phenylalanine, 99%
D-Phenylalanine, 99%
D-Phenylalanine, 99%
Thermo Scientific Chemicals
D-Phenylalanine, 99%
CAS: 673-06-3 | C9H11NO2 | 165.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number ALFA10572.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS14024-18-1
IUPAC Nameiron(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC15H21FeO6
InChI KeyAQBLLJNPHDIAPN-LNTINUHCSA-K
SMILES[Fe+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Volatile content: 0.30%, max
Formpowder
Appearance (Color)Red
Insoluble material0.30 wt%, max
Melting Point170 to 190 deg C
It is an essential amino acid with anti-depressant and analgesic activities. pharmaceutical intermediates, mainly used to treat, prevent osteoporosis, cardiovascular, diabetes, arterioscleros.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an essential amino acid with anti-depressant and analgesic activities. pharmaceutical intermediates, mainly used to treat, prevent osteoporosis, cardiovascular, diabetes, arterioscleros.
Solubility
Soluble in water (30 mg/ml). Slightly soluble in methanol, ethanol, DMSO.
Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. D W Christianson; S Mangani; G Shoham and W N Lipscomb. Binding of D-phenylalanine and D-tyrosine to carboxypeptidase A. J Biol Chem. 2004, 279, 26046-26051.
  2. J. Huang; V.M. Egan; H. Guo; J.-Y. Yoon; A.L. Briseno; I.E. Rauda; R.L.Garrell; C.M. Knobler; F. Zhou; R.B. Kaner. Enantioselective Discrimination of D- and L-Phenylalanine by Chiral Polyaniline Thin Films. Adv. Mater. 2003, 15, (14) 223-228.