Iodomethane, 99%, stab. with copper, Thermo Scientific Chemicals
Iodomethane, 99%, stab. with copper, Thermo Scientific Chemicals
Iodomethane, 99%, stab. with copper, Thermo Scientific Chemicals
Iodomethane, 99%, stab. with copper, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodomethane, 99%, stab. with copper, Thermo Scientific Chemicals
CAS: 74-88-4 | CH3I | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number ALFA10559.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialIodomethane
Name NoteStabilized with copper
CAS74-88-4
Health Hazard 1H301+H331-H312-H315-H319-H335-H351
Health Hazard 2GHS H Statement
H301-H331-H351-H312-H315-H335
Toxic if swallowed.
Toxic if inhaled.
Suspected of causing cancer.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
View more
Iodomethane is a methylating agent used for alkylating carbon, oxygen, sulfur, nitrogen and phosphorus nucleophiles. For example, phenol reacts with methyl iodide to give anisole. In the Monsanto process, the insitu formed iodomethane reacts with carbon monoxide in the presence of a rhodium complex to give acetyl iodide, which on hydrolysis yields acetic acid. It reacts with magnesium to form the Grignard reagent, methylmagnesium iodide used in organic synthesis. It finds applications as an intermediate in the manufacture of pharmaceuticals and in phase transfer catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Iodomethane is a methylating agent used for alkylating carbon, oxygen, sulfur, nitrogen and phosphorus nucleophiles. For example, phenol reacts with methyl iodide to give anisole. In the Monsanto process, the insitu formed iodomethane reacts with carbon monoxide in the presence of a rhodium complex to give acetyl iodide, which on hydrolysis yields acetic acid. It reacts with magnesium to form the Grignard reagent, methylmagnesium iodide used in organic synthesis. It finds applications as an intermediate in the manufacture of pharmaceuticals and in phase transfer catalysts.
Solubility
Miscible with water.
Notes
Light and moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases and oxygen.
RUO – Research Use Only
General References:
  1. Erk, B.; Boll, R., Trippel, S.; Anielski, D.; Foucar, L.; Rudek, B.; Rudenko, A. Imaging charge transfer in iodomethane upon x-ray photoabsorption. Science 2014, 345 (6194), 288-291.
  2. Dong, Y.; Brooks, J. D.; Chen, T. L.; Mullins, D. R.; Cox, D. F. Reactions of methyl groups on a non-reducible metal oxide: The reaction of iodomethane on stoichiometric alpha-Cr 2 O 3 (0001). Surf. Sci. 2015, 641, 148-153.