Indole-3-acetic acid, 98+%, Thermo Scientific Chemicals
Indole-3-acetic acid, 98+%, Thermo Scientific Chemicals
Indole-3-acetic acid, 98+%, Thermo Scientific Chemicals
Indole-3-acetic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Indole-3-acetic acid, 98+%, Thermo Scientific Chemicals
CAS: 87-51-4 | C10H9NO2 | 175.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFA10556.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7601-54-9
IUPAC Nametrisodium phosphate
Molecular FormulaNa3O4P
InChI KeyRYFMWSXOAZQYPI-UHFFFAOYSA-K
SMILES[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HCl>=96.0 %
Appearance (Color)White
It is applied as plant growth hormone. is an inducer of plant cell elongation and division shown to cause uncontrolled growth.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied as plant growth hormone. is an inducer of plant cell elongation and division shown to cause uncontrolled growth.
Solubility
Soluble in ethanol (50 mg/ml), methanol, DMSO, and chloroform (sparingly). Insoluble in water.
Notes
Light sensitive. Store at -20°C. ncompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Cheryl L. Patten; Bernard R. Glick. Bacterial biosynthesis of indole-3-acetic acid. Can. J. Microbiol. 1996, 42, (3), 207-220.
  2. J. RIGAUD, A. PUPPO. Indole-3-acetic Acid Catabolism by Soybean Bacteroids. Microbiology. 1975, 88, 223-228.