p-Xylene, 99%, Thermo Scientific Chemicals
p-Xylene, 99%, Thermo Scientific Chemicals
p-Xylene, 99%, Thermo Scientific Chemicals
p-Xylene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Xylene, 99%, Thermo Scientific Chemicals
CAS: 106-42-3 | C8H10 | 106.17 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10534.AP500 mL
Catalog number ALFA10534.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or Materialp-Xylene
Melting Point12°C to 13°C
CAS106-42-3
Health Hazard 1H226-H304-H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H226-H312-H332-H315
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
View more
p-Xylene is used as a precursor in the production of benzoic, isophthalic, tetraphillic acids and dimethyle esters, which are used in the manufacture of polyester. It acts as an intermediate in plastic and rubber products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Xylene is used as a precursor in the production of benzoic, isophthalic, tetraphillic acids and dimethyle esters, which are used in the manufacture of polyester. It acts as an intermediate in plastic and rubber products.
Solubility
Miscible with alcohol, ether, acetone, benzene and chloroform. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Wijaya, Y. P.; Suh, D. J.; Jae, J. Production of renewable p-xylene from 2, 5-dimethylfuran via Diels-Alder cycloaddition and dehydrative aromatization reactions over silica- alumina aerogel catalysts. Catal. Commun. 2015, 70, 3716-3722.
  2. Ahn, J. H.; Kolvenbach, R.; Gutiérrez, O. Y.; Al-Khattaf, S. S.; Jentys, A.; Lercher, J. A. Tailoring p-xylene selectivity in toluene methylation on medium pore-size zeolites. Microporous Mesoporous Mater. 2015, 210, 52-59.