Ammonium peroxydisulfate, 98%, Thermo Scientific Chemicals
Ammonium peroxydisulfate, 98%, Thermo Scientific Chemicals
Ammonium peroxydisulfate, 98%, Thermo Scientific Chemicals
Ammonium peroxydisulfate, 98%, Thermo Scientific Chemicals
Ammonium peroxydisulfate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium peroxydisulfate, 98%, Thermo Scientific Chemicals
CAS: 7727-54-0 | H8N2O8S2 | 228.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
5000 g
Catalog number ALFA10533.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium peroxydisulfate
Melting Point120°C (decomposition)
CAS7727-54-0
Health Hazard 1H272-H302-H315-H317-H319-H334-H335
Health Hazard 2GHS H Statement
H272-H301-H334-H315-H319-H317-H335
May intensify fire
oxidizer.
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
View more
Ammonium peroxydisulfate is widely utilized as an oxidizing agent and radical initiators in the polymerization of certain alkenes. It is actively involved in in the synthesis of commercially important polymers like styrene-butadiene rubber and polytetrafluoroethylene. It combines with tetramethylethylenediamine to catalyze the polymerization of acrylamide. Due to its oxidizing properties, it can be employed to etch copper on printed circuit boards which is an alternative to ferric chloride solution. Also, it acts as oxidants in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ammonium peroxydisulfate is widely utilized as an oxidizing agent and radical initiators in the polymerization of certain alkenes. It is actively involved in in the synthesis of commercially important polymers like styrene-butadiene rubber and polytetrafluoroethylene. It combines with tetramethylethylenediamine to catalyze the polymerization of acrylamide. Due to its oxidizing properties, it can be employed to etch copper on printed circuit boards which is an alternative to ferric chloride solution. Also, it acts as oxidants in organic synthesis.
Solubility
Freely soluble in water.
Notes
Aqueous solution decomposes slowly at room temperature.Moisture sensitive. Incompatible with strong reducing agents, organic materials and powdered metals.
RUO – Research Use Only
General References:
  1. In combination with AgNO3 and Cu(OAc)2, promotes a carbamoyl radical process for the conversion of oxalic acid monoamides to isocyanates: J. Org. Chem., 60, 5430 (1995).
  2. Hosseini, S. H. Kinetics study of copolymerization of 2-anilinoethanol onto chitosan by ammonium peroxydisulfate as a initiator. Int. J. Phys. Sci. 2015, 10 (4), 142-154.
  3. Gopalakrishnan, k.; Sultan, S.; Govindaraj, A.; Rao, C. N. R. Supercapacitors based on composites of PANI with nanosheets of nitrogen-doped RGO, BC1.5N, MoS2and WS2. Nano Energy 2015, 12 52-58.