Glycylglycine, 99%
Glycylglycine, 99%
Glycylglycine, 99%
Thermo Scientific Chemicals
Glycylglycine, 99%
CAS: 556-50-3 | C4H8N2O3 | 132.119 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10523.22100 g
Catalog number ALFA10523.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGlycylglycine
Melting Point∼220°C (decomposition)
CAS556-50-3
Health Hazard 1H319
Health Hazard 2GHS H Statement
H319
Causes serious eye irritation.
View more
Glycylglycine is as a buffering reagent and also used in biochemical research. It is the simplest of all dipeptides and is used as a starting template for preparation of more complex peptides. It is also employed as a practical buffer with a buffering range of 7.5 - 8.9 (at 25°C). Valuable agent for experiments investigating peptide transport and absorption.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glycylglycine is as a buffering reagent and also used in biochemical research. It is the simplest of all dipeptides and is used as a starting template for preparation of more complex peptides. It is also employed as a practical buffer with a buffering range of 7.5 - 8.9 (at 25°C). Valuable agent for experiments investigating peptide transport and absorption.
Solubility
Soluble in water (132 g/L) at 20°C, and ethanol (slightly solube).
Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Dr. David A.Leigh, Aden Murphy, Dr. John P.Smart Alexandra M.Z.Slawin. Glycylglycine Rotaxanes—The Hydrogen Bond Directed Assembly of Synthetic Peptide Rotaxanes . Angewandte Chemie International Edition in English. 1997, 36, (7),728-732
  2. Noriko Takuwa; Tadahito Shimada; Hisashi Matsumoto; Takeshi Hoshi. Proton-coupled transport of glycylglycine in rabbit renal brush-border membrane vesicles. Biochimica et Biophysica Acta (BBA) - Biomembranes. 1985, 814, (1),186-190