1-Bromopropane, 99%, Thermo Scientific Chemicals
1-Bromopropane, 99%, Thermo Scientific Chemicals
1-Bromopropane, 99%, Thermo Scientific Chemicals
1-Bromopropane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromopropane, 99%, Thermo Scientific Chemicals
CAS: 106-94-5 | C3H7Br | 122.993 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 mL
250 mL
Catalog number ALFA10461.AU
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 mL
Request bulk or custom format
Chemical Identifiers
CAS4249-10-9
IUPAC Name1,2,3,4-tetramethylcyclopenta-1,3-diene
Molecular FormulaC9H14
InChI KeyVNPQQEYMXYCAEZ-UHFFFAOYSA-N
SMILESCC1=C(C)C(C)=C(C)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow to orange
FormLiquid
Assay (GC)≥80.0% (mixture of isomers)
Refractive Index1.4700-1.4760 @ 20°C
1-Bromopropane is used as a solvent for adhesives, degreasing plastics and optics. It is used in the production of asphalt, synthetic fiber and aerosols. It is employed for cleaning the metal surfaces, electronic circuit boards, and multi-layered OLEDs (Organic Light Emitting Diodes). It serves to replace perchloroethylene as a dry cleaning solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromopropane is used as a solvent for adhesives, degreasing plastics and optics. It is used in the production of asphalt, synthetic fiber and aerosols. It is employed for cleaning the metal surfaces, electronic circuit boards, and multi-layered OLEDs (Organic Light Emitting Diodes). It serves to replace perchloroethylene as a dry cleaning solvent.
Solubility
Miscible with ethanol, water and diethyl ether.
Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
  1. Garner, C. E.; Liang, S.; Yin, L.; Yu, X. Physiologically based pharmacokinetic modeling for 1-bromopropane in F344 rats using gas uptake inhalation experiments. Toxicol. Sci. 2015, 145 (1), 23-36.
  2. Frasch, H. F.; Dotson, G. S.; Barbero, A. M. In vitro Human Epidermal Penetration of 1-Bromopropane. J. Toxicol. Environ. Health A 2011, 74 (19), 1249-1260.