L-Histidine, 98+%
L-Histidine, 98+%
L-Histidine, 98+%
Thermo Scientific Chemicals
L-Histidine, 98+%
CAS: 71-00-1 | C6H9N3O2 | 155.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number ALFA10413.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL-Histidine
Melting Point∼281°C (decomposition)
CAS71-00-1
pH7.0 to 8
Recommended StorageAmbient temperatures
View more
L-Histidine acts as a precursor to histamine and a component of carnosine. It is also used in medicine, feed additive, biochemical research, dietary supplement. It is used in a nutrition enhancer, is the amino acid fluid infusion. It can be used in the treatment of gastric ulcer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Histidine acts as a precursor to histamine and a component of carnosine. It is also used in medicine, feed additive, biochemical research, dietary supplement. It is used in a nutrition enhancer, is the amino acid fluid infusion. It can be used in the treatment of gastric ulcer.
Solubility
Soluble in water (15 mg/ml at 20°C).
Notes
Store at 4°C.Incompatible with oxidizing agents.
RUO – Research Use Only
JJ Madden; EL McGandy; NC Seeman. The crystal structure of the orthorhombic form of L-(+)-histidine. Acta Cryst. 1972, B28, 2377-2382.