Citric acid, 99+%, Thermo Scientific Chemicals
Citric acid, 99+%, Thermo Scientific Chemicals
Citric acid, 99+%, Thermo Scientific Chemicals
Citric acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citric acid, 99+%, Thermo Scientific Chemicals
CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
Catalog number ALFA10395.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Chemical Identifiers
CAS106-48-9
IUPAC Name4-chlorophenol
Molecular FormulaC6H5ClO
InChI KeyWXNZTHHGJRFXKQ-UHFFFAOYSA-N
SMILESOC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or crystalline powder or fused/lumpy solid
Assay (Silylated GC)≥98.5% (UK-sourced material)
Melting Point33-46?C
CommentMaterial sourced in UK and US
View more
Citric acid is the most widely used organic acid and pH-control agent in foods, beverages, pharmaceuticals and technical applications. Used in pharmaceutical preparations, especially in effervescent tablets. It is used as a natural preservative.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Citric acid is the most widely used organic acid and pH-control agent in foods, beverages, pharmaceuticals and technical applications. Used in pharmaceutical preparations, especially in effervescent tablets. It is used as a natural preservative.
Solubility
Soluble in water, alcohol, ether, dimethylsufoxide and ethylacetate.
Notes
Hygroscopic. Incompatible with alkalies, bases, metal nitrates and oxidizing agents.
RUO – Research Use Only
General References:
  1. Pan, K.; Zhong, Q. Improving Clarity and Stability of Skim Milk Powder Dispersions by Dissociation of Casein Micelles at pH 11.0 and Acidification with Citric Acid. Agric. Food Chem. 2013, 61 (38), 9260-9268.
  2. Kolah, A. K.; Asthana, N. S.; Vu, D. T.; Carl T. Lira.; Miller, D. J. Reaction Kinetics of the Catalytic Esterification of Citric Acid with Ethanol. Ind. Eng. Chem. Res. 2007, 46 (10), 3180-3187.
  3. Gogoi, P.; Boruah, M.; Sharma, S.; Dolui, S. K. Blends of Epoxidized Alkyd Resins Based on Jatropha Oil and the Epoxidized Oil Cured with Aqueous Citic Acid Solution: A Green Technology Approach. ACS Sustainable Chem. Eng. 2015, 3 (2), 261-268.