Dodecanedioic acid, 99%, Thermo Scientific Chemicals
Dodecanedioic acid, 99%, Thermo Scientific Chemicals
Dodecanedioic acid, 99%, Thermo Scientific Chemicals
Dodecanedioic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dodecanedioic acid, 99%, Thermo Scientific Chemicals
CAS: 693-23-2 | C12H22O4 | 230.304 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFA10387.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS3886-70-2
IUPAC Name(1R)-1-(naphthalen-1-yl)ethan-1-aminium
Molecular FormulaC12H14N
InChI KeyRTCUCQWIICFPOD-SECBINFHSA-O
SMILESC[C@@H]([NH3+])C1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Refractive index1.6215 to 1.6245 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
View more
Dodecanedioic acid is used as an intermediate for plasticizers, lubricants and adhesives. It is also used in antiseptics, top-grade coatings, painting materials, corrosion inhibitor and surfactant. It finds application in engineering plastics such as nylon 612.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dodecanedioic acid is used as an intermediate for plasticizers, lubricants and adhesives. It is also used in antiseptics, top-grade coatings, painting materials, corrosion inhibitor and surfactant. It finds application in engineering plastics such as nylon 612.
Solubility
Soluble in water, ethanol, hot methanol, hot toluene and hot acetic acid.
Notes
Incompatible with strong oxidizing agents and reducing agents.
RUO – Research Use Only
General References:
  1. Alhendawi, H.; Brunet, E.; Juanes, O.; Hammouda, H.; Idhair, S.; Payán, E. R.; de Victoria Rodríguez, M. New soft porous frameworks based on lambda-zirconium phosphate and aliphatic dicarboxylates: Synthesis and structural characterization. J. Phys. Chem. Solids 2015, 86, 95-100.
  2. Elbasuney, S. Surface engineering of layered double hydroxide (LDH) nanoparticles for polymer flame retardancy. Powder Technol. 2015, 277, 63-73.