N,N-Dimethyl-p-phenylenediamine dihydrochloride, 98%, Thermo Scientific Chemicals
N,N-Dimethyl-p-phenylenediamine dihydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethyl-p-phenylenediamine dihydrochloride, 98%, Thermo Scientific Chemicals
CAS: 536-46-9 | C8H14Cl2N2 | 209.11 g/mol
Have Questions?
Catalog NumberdimQuantity
ALFA10343.14
Catalog number ALFA10343.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight209.12
Unit Size
It is a stain used in microscopy. N,N-Dimethyl-p-phenylenediamine dihydrochloride is also suitable for use in the peroxidase test. It is a compound that is normally used to measure the antioxidant potential.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a stain used in microscopy. N,N-Dimethyl-p-phenylenediamine dihydrochloride is also suitable for use in the peroxidase test. It is a compound that is normally used to measure the antioxidant potential.
Solubility
Soluble in water
Notes
Store away from oxidizing agents, humidity and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from light. Store under inert gas.
RUO – Research Use Only
General References:
  1. Kazuo Hirayama and Nobuyuki Unohara. Spectrophotometric catalytic determination of an ultratrace amount of iron(III) in water based on the oxidation of N,N-dimethyl-p-phenylenediamine by hydrogen peroxide.Anal. Chem.,1988,60(23), 2573-2577.
  2. Leland L. Smith and Freddie L. Hill. Detection of sterol hydroperoxides on thin-layer chromatoplates by means of the wurster dyes.J. Chromatogr. A,1972,66(1), 101-109.