Bisphenol A, 97+%, Thermo Scientific Chemicals
Bisphenol A, 97+%, Thermo Scientific Chemicals
Bisphenol A, 97+%, Thermo Scientific Chemicals
Bisphenol A, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bisphenol A, 97+%, Thermo Scientific Chemicals
CAS: 80-05-7 | C15H16O2 | 228.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFA10324.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS2627-86-3
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.5 %
Refractive index1.5250 to 1.5270 (20°C, 589 nm)
Specific optical rotation-39° to -41° (20°C, 589 nm) (neat)
Appearance (Color)Clear colorless to light yellow
View more
Bisphenol A is an important monomer used in the production of polycarbonate plastics which is utilized to make various consumer goods like water bottles, sports equipment, medical devices, eyeglass lenses, house hold electronics, compact discs (CDs) and digital versatile discs (DVDs). Bisphenol A containing epoxy resins are used to make lining in water pipes. It is used as a precursor to prepare flame retardant tetrabromobispheonl A, polysulfones and polyether ketones, which are antioxidants in some plasticizers. It acts as a polymerization inhibitor in poly vinyl chloride (PVC).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bisphenol A is an important monomer used in the production of polycarbonate plastics which is utilized to make various consumer goods like water bottles, sports equipment, medical devices, eyeglass lenses, house hold electronics, compact discs (CDs) and digital versatile discs (DVDs). Bisphenol A containing epoxy resins are used to make lining in water pipes. It is used as a precursor to prepare flame retardant tetrabromobispheonl A, polysulfones and polyether ketones, which are antioxidants in some plasticizers. It acts as a polymerization inhibitor in poly vinyl chloride (PVC).
Solubility
Soluble in aqueous alkaline solution, acetic acid, alcohol and acetone. Slightly soluble in carbon tetrachloride. It has poor solubility in water.
Notes
Incompatible with acid anhydrides, acid chlorides, strong bases and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Myridakis, A.; Balaska, E.; Gkaitatzi, C.; Kouvarakis, A.; Stephanou, E. G. Determination and separation of bisphenol A, phthalate metabolites and structural isomers of parabens in human urine with conventional high-pressure liquid chromatography combined with electrospray ionisation tandem mass spectrometry. Anal. Bioanal. Chem. 2015, 407 (9), 2509-2518.
  2. Zhao, J.; Ma, Y.; Hou, X.; Li, L.; Zheng, P.; Li, C. A novel ionic liquid synthesis, electrochemical polymerization, and sensing performance toward bisphenol A. J. Solid State Electrochem. 2015, 19 (6), 1571-1578.