5-Chloro-2-pentanone, 94%, Thermo Scientific Chemicals
5-Chloro-2-pentanone, 94%, Thermo Scientific Chemicals
5-Chloro-2-pentanone, 94%, Thermo Scientific Chemicals
5-Chloro-2-pentanone, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Chloro-2-pentanone, 94%, Thermo Scientific Chemicals
CAS: 5891-21-4 | C5H9ClO | 120.576 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10269.22100 g
Catalog number ALFA10269.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material5-Chloro-2-pentanone
CAS5891-21-4
Health Hazard 1H226-H302-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
View more
5-Chloro-2-pentanone used in the preparation of destructible surfactants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Chloro-2-pentanone used in the preparation of destructible surfactants.
Solubility
Soluble in chloroform and methanol.
Notes
keep container tightly closed. Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents
RUO – Research Use Only
General References:
  1. Hideo Kise; Toru Sato; Tsutomu Yasuoka; Manabu Seno; Teruzo Asahara. Synthesis of ocimenol and related compounds from 5-chloro-2-pentanone. J. Org. Chem. 1979, 44, (24), 4454-4456.
  2. David A. Jaeger; Moira R. Frey. Preparation and characterization of destructible surfactants. J. Org. Chem. 1982, 47, (2), 311-315.