Ammonium oxalate monohydrate, 98%, Thermo Scientific Chemicals
Ammonium oxalate monohydrate, 98%, Thermo Scientific Chemicals
Ammonium oxalate monohydrate, 98%, Thermo Scientific Chemicals
Ammonium oxalate monohydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium oxalate monohydrate, 98%, Thermo Scientific Chemicals
CAS: 6009-70-7 | C2H10N2O5 | 142.111 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFA10263.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS91983-26-5
IUPAC Name[4-(cyanomethyl)phenyl]boronic acid
Molecular FormulaC8H8BNO2
InChI KeyYKVMTTIYBSVPEQ-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(CC#N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (HPLC)≥94.0%
Appearance (Color)White to cream to yellow to green to orange to pale brown
Assay (Aqueous acid-base Titration)≥94.0%
Proton NMRConforms to structure
Ammonium oxalate monohydrate is widely utilized as a buffering reagent and a dispersant to determine the surface interaction of platelets. It finds an application to study its acute poisoning effect on sheep and to investigate the formation of oxalate monoalkylamide in the human lens. It is also used in the detection and determination of calcium, lead, fluoride and rare earth metals. It is employed as chelators and forms complexes with metals. It acts as reducing agent in gold extraction and is an active ingredient of ferrous metal surface polishing fluid. It is a promoting agent in production of cobalt oxide and nickel oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ammonium oxalate monohydrate is widely utilized as a buffering reagent and a dispersant to determine the surface interaction of platelets. It finds an application to study its acute poisoning effect on sheep and to investigate the formation of oxalate monoalkylamide in the human lens. It is also used in the detection and determination of calcium, lead, fluoride and rare earth metals. It is employed as chelators and forms complexes with metals. It acts as reducing agent in gold extraction and is an active ingredient of ferrous metal surface polishing fluid. It is a promoting agent in production of cobalt oxide and nickel oxide.
Solubility
Soluble in water. Slightly soluble in alcohol.
Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
General References:
  1. Kripal, R.; Yadav, A. K. Theoretical calculation of zero field splitting parameters of Cr3+doped ammonium oxalate monohydrate. Physica B. 2015, 466, 16-18.
  2. Qiao, Y.; Wang, K.; Yuan, H.; Yang, K.; Zou, B. Negative Linear Compressibility in Organic Mineral Ammonium Oxalate Monohydrate with Hydrogen Bonding Wine-Rack Motifs. J. Phys. Chem. Lett. 2015, 6 (14), 2755-2760.