4-Fluorobenzyl bromide, 97%, Thermo Scientific Chemicals
4-Fluorobenzyl bromide, 97%, Thermo Scientific Chemicals
4-Fluorobenzyl bromide, 97%, Thermo Scientific Chemicals
4-Fluorobenzyl bromide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Fluorobenzyl bromide, 97%, Thermo Scientific Chemicals
CAS: 459-46-1 | C7H6BrF | 189.03 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10208.0910 g
Catalog number ALFA10208.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS81090-53-1
IUPAC Name4-bromo-N-(4-bromophenyl)-N-phenylaniline
Molecular FormulaC18H13Br2N
InChI KeyKIGVOJUDEQXKII-UHFFFAOYSA-N
SMILESBrC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%