Benzoin, 99%, Thermo Scientific Chemicals
Benzoin, 99%, Thermo Scientific Chemicals
Benzoin, 99%, Thermo Scientific Chemicals
Benzoin, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzoin, 99%, Thermo Scientific Chemicals
CAS: 119-53-9 | C14H12O2 | 212.25 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10188.30250 g
Catalog number ALFA10188.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Comment0.023 - 0.028mm thick
FormFoil
Appearance (Color)Copper
Total Metal Impurities0.001% max.
Benzoin is used as a flavor, antiseptic and photopolymerization catalyst. It is also used as a raw material in organic syntheses. It serves as a precursor to benzil, which is a photoinitiator. Further, it is used as degassing agent for powder coatings. In addition to this, it is used as an additive and in powder coating to remove the pinhole phenomenon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzoin is used as a flavor, antiseptic and photopolymerization catalyst. It is also used as a raw material in organic syntheses. It serves as a precursor to benzil, which is a photoinitiator. Further, it is used as degassing agent for powder coatings. In addition to this, it is used as an additive and in powder coating to remove the pinhole phenomenon.
Solubility
Soluble in chlorine. Slightly soluble in water, ethanol and ether.
Notes
Moisture sensitive. Incompatible with acids, acid anhydrides, acid chlorides and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Exclusive C-alkylation occurs with an alkyl bromide and NaOH in DMSO: Liebigs Ann. Chem., 735, 56 (1970). Borohydride reduction and periodate cleavage of the product lead to a useful synthesis of aryl ketones, with benzoin functioning as a benzoyl anion equivalent: Synthesis, 268 (1975):
  2. This has been extended to include alkylation with other electrophiles including primary and secondary alkyl iodides and Michael acceptors, and the route shortened by direct cleavage of the alkylated benzoin with hydrogen peroxide: Synthesis, 36 (1980).
  3. Zeng, H.; Wang, K.; Tian, Y.; Niu, Y.; Greene, L.; Hu, Z.; Lee, J. K. Reprint of The benzoin condensation: Charge tagging of the catalyst allows for tracking by mass spectrometry. Int. J. Mass Spectrom. 2015, 378, 169-174.
  4. Alamgir, M.; Khuhawar, M. Y.; Memon, S. Q.; Hayat, A.; Zounr, R. A. Spectrofluorimetric analysis of famotidine in pharmaceutical preparations and biological fluids by derivatization with benzoin. Spectrochim. Acta, Part A 2015, 134, 449-452.