Cadmium sulfate hydrate, 98%, Thermo Scientific Chemicals
Cadmium sulfate hydrate, 98%, Thermo Scientific Chemicals
Cadmium sulfate hydrate, 98%, Thermo Scientific Chemicals
Cadmium sulfate hydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cadmium sulfate hydrate, 98%, Thermo Scientific Chemicals
CAS: 15244-35-6 | CdO4S | 208.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFA10168.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS137-43-9
IUPAC Namebromocyclopentane
Molecular FormulaC5H9Br
InChI KeyBRTFVKHPEHKBQF-UHFFFAOYSA-N
SMILESBrC1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4870 to 1.4910 (20°C, 589 nm)
GC>=97.5 %
Cadmium sulfate hydrate is used for formulating screens or for optimization. It is also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cadmium sulfate hydrate is used for formulating screens or for optimization. It is also used as pharmaceutical intermediates.
Solubility
Soluble in water (1130g/L ).
Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. M. N. Papadopoulos; W. F. Giauque. Cadmium Sulfate and its Hydrates. Heat Capacities and Heats of Hydration. Application of the Third Law of Thermodynamics1. J. Am. Chem. Soc. 1955, 77,(10), 2740-2744.
  2. P. DuBois Murphy; B. C. Gerstein. Cadmium Sulfate and its Hydrates. Principal components of the cadmium-113 shielding tensors in cadmium sulfate hydrates: a nuclear magnetic resonance study of cadmium coordinated with oxygen. J. Am. Chem. Soc. 1981, 103,(12), 3282-3286.