Catechol, 99%, Thermo Scientific Chemicals
Catechol, 99%, Thermo Scientific Chemicals
Catechol, 99%, Thermo Scientific Chemicals
Catechol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Catechol, 99%, Thermo Scientific Chemicals
CAS: 120-80-9 | C6H6O2 | 110.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
Catalog number ALFA10164.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Chemical Identifiers
CAS10361-37-2
IUPAC Namebarium(2+) dichloride
Molecular FormulaBaCl2
InChI KeyWDIHJSXYQDMJHN-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Ba++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder, may contain small lumps
Total Metal Impurities≤0.002% (20 ppm)
Catechol is used as a developer for photographic films, dyes and an intermediate for antioxidants in rubber and lubricating oils. It is used in polymerization inhibitors and in pharmaceuticals. It is a building block in organic synthesis and involved in the preparation of flavors and fragrances. It is used as a replacement for sandalwood oil, which is prepared from catechol through guaiacol and camphor. Further, it is a precursor for the preparation of vanillin from guaiacol, which is obtained by methylation reaction with catechol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Catechol is used as a developer for photographic films, dyes and an intermediate for antioxidants in rubber and lubricating oils. It is used in polymerization inhibitors and in pharmaceuticals. It is a building block in organic synthesis and involved in the preparation of flavors and fragrances. It is used as a replacement for sandalwood oil, which is prepared from catechol through guaiacol and camphor. Further, it is a precursor for the preparation of vanillin from guaiacol, which is obtained by methylation reaction with catechol.
Solubility
Soluble in water, pyridine, chloroform, benzene, carbon tetrachloride, ether and acetate.
Notes
Air and light sensitive. Incompatible with oxidizing agents, aluminum, bases, acid chlorides and acid anhydrides.
RUO – Research Use Only
General References:
  1. Ring-opening with O2 in the presence of CuCl, pyridine and methanol gives the monoester of cis,cis-muconic acid: Org. Synth. Coll., 8, 490 (1993).
  2. In the presence of catechol, carboxylic acids are reduced to alcohols by NaBH4: Tetrahedron, 48, 371 (1992).
  3. For an example of the use of catechol in the synthesis of crown ethers, see: Org. Synth. Coll., 6, 395 (1988).
  4. Jastrzebski, R.; van den Berg, E. J.; Weckhuysen, B. M.; Bruijnincx, P. C. A. Sustainable production of dimethyl adipate by non-heme iron(III) catalysed oxidative cleavage of catechol. Catal. Sci. Technol. 2015, 5 (4), 2103-2109.
  5. Zhong, M.; Dai, Y.; Fan, L.; Lu, X.; Kan, X. A novel substitution-sensing for hydroquinone and catechol based on a poly(3-aminophenylboronic acid)/MWCNTs modified electrode. Analyst 2015, 140 (17), 6047-6053.