2-Iodoanisole, 99%, Thermo Scientific Chemicals
2-Iodoanisole, 99%, Thermo Scientific Chemicals
2-Iodoanisole, 99%, Thermo Scientific Chemicals
2-Iodoanisole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Iodoanisole, 99%, Thermo Scientific Chemicals
CAS: 529-28-2 | C7H7IO | 234.036 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10150.1425 g
Catalog number ALFA10150.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS128427-10-1
IUPAC Nametert-butyl N-[(2S)-1,4-dihydroxybutan-2-yl]carbamate
Molecular FormulaC9H19NO4
InChI KeyKLRRFBSWOIUAHZ-ZETCQYMHSA-N
SMILESCC(C)(C)OC(=O)N[C@H](CO)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White
Appearance (Form)Crystalline powder
Melting point65°C to 69°C
GC>=96.0 %
View more
2-Iodoanisole has been used in palladium/copper-catalyzed synthesis of o-(1-alkynyl)anisoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Iodoanisole has been used in palladium/copper-catalyzed synthesis of o-(1-alkynyl)anisoles.
Solubility
Miscible with alcohol, chloroform and diethyl ether. Insoluble in water.
Notes
Light Sensitive, store in dark. Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Dawei Yue et. al.Synthesis of 2,3-disubstituted benzo[b]furans by the palladium-catalyzed coupling of o-iodoanisoles and terminal alkynes, followed by electrophilic cyclization. Journal of Organic Chemistry. 2005, 70 (25), 10292-10296 .
  2. Adam Morel et. al. Palladium-catalyzed asymmetric Heck arylation of 2,3-dihydrofuran--effect of prolinate salts. Dalton Transactions. 2013, 42 (4), 1215-1222.