Phosphorus, AAS standard solution, Specpure™ P 1000μg/mL
Phosphorus, AAS standard solution, Specpure™ P 1000μg/mL
Phosphorus, AAS standard solution, Specpure™ P 1000μg/mL
Phosphorus, AAS standard solution, Specpure™ P 1000μg/mL
Thermo Scientific Chemicals
Phosphorus, AAS standard solution, Specpure™ P 1000μg/mL
CAS: 7723-14-0 | P | 30.97 g/mol
Have Questions?
Catalog NumberQuantity
ALF089887.AP500 mL
Catalog number ALF089887.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialPhosphorus
CAS7723-14-0
Quantity500 mL
Health Hazard 1H314 : Causes severe skin burns and eye damage.
CAS7697-37-2