Phosphorus, AAS standard solution, Specpure™ P 1000μg/mL
Phosphorus, AAS standard solution, Specpure™ P 1000μg/mL
Phosphorus, AAS standard solution, Specpure™ P 1000μg/mL
Phosphorus, AAS standard solution, Specpure™ P 1000μg/mL
Thermo Scientific Chemicals
Phosphorus, AAS standard solution, Specpure™ P 1000μg/mL
CAS: 7723-14-0 | P | 30.97 g/mol
Have Questions?
Catalog NumberQuantity
ALF089887.AE100 mL
Catalog number ALF089887.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS6381-92-6
IUPAC Namedisodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Molecular FormulaC10H18N2Na2O10
InChI KeyOVBJJZOQPCKUOR-UHFFFAOYSA-L
SMILESO.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Protease activityPasses test
DNase(exo): Passes test
RNasePasses test
DNase(endo): Passes test
pH8.0 ? 0.5