Calcium phosphate (pyro), 96% min, Thermo Scientific Chemicals
Calcium phosphate (pyro), 96% min, Thermo Scientific Chemicals
Calcium phosphate (pyro), 96% min, Thermo Scientific Chemicals
Calcium phosphate (pyro), 96% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Calcium phosphate (pyro), 96% min, Thermo Scientific Chemicals
CAS: 7790-76-3 | Ca2O7P2 | 254.10 g/mol
Have Questions?
Catalog NumberQuantity
ALF089836.A11 kg
Catalog number ALF089836.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialCalcium phosphate (pyro)
Melting Point1230°C
CAS7790-76-3
ColorWhite
Health Hazard 1H315-H319-H335
View more
Calcium phosphate is used in the production of phosphoric acid and fertilizers. It is used in baking as the acid in a leavening agent. It is also used in cheese products. This compound has also been used in gene transfection of cells. It is also being used in the development of contrast agents.