Bismuth(III) chloride, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals
Bismuth(III) chloride, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals
Bismuth(III) chloride, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals
Bismuth(III) chloride, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bismuth(III) chloride, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 7787-60-2 | BiCl3 | 315.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Catalog number ALF089485.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight315.34
Unit Size
It is used as a catalyst in organic chemistry. It is a good source of bismuth and is used to make other bismuth compounds. Also used as a catalyst and to make other bismuth salts.