Kieselguhr, Thermo Scientific Chemicals
Kieselguhr, Thermo Scientific Chemicals
Kieselguhr, Thermo Scientific Chemicals
Kieselguhr, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Kieselguhr, Thermo Scientific Chemicals
CAS: 61790-53-2 | O2Si | 60.08 g/mol
Have Questions?
Catalog NumberQuantity
ALF089381.A11 kg
Catalog number ALF089381.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS75-31-0
IUPAC Namepropan-2-amine
Molecular FormulaC3H9N
InChI KeyJJWLVOIRVHMVIS-UHFFFAOYSA-N
SMILESCC(C)N
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.3725-1.3755 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥99.0%
FormLiquid
Kieselguhr is used in used as a filtration aid, mild abrasive in products including metal polishes and toothpaste, absorbent for liquids, matting agent for coatings, reinforcing filler in plastics and rubber, anti-block in plastic films, porous support for chemical catalysts, cat litter, activator in blood clotting studies, a stabilizing component of dynamite, and a thermal insulator.