Ethylenediaminetetraacetic acid, iron(III) monosodium salt, Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, iron(III) monosodium salt, Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, iron(III) monosodium salt, Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, iron(III) monosodium salt, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, iron(III) monosodium salt, Thermo Scientific Chemicals
CAS: 15708-41-5 | C10H12FeN2NaO8 | 367.047 g/mol
Have Questions?
Catalog NumberQuantity
ALF088995.22100 g
Catalog number ALF088995.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS12070-12-1
IUPAC Namemethanidylidynetungstenylium
Molecular FormulaCW
InChI KeyUONOETXJSWQNOL-UHFFFAOYSA-N
SMILES[C-]#[W+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Assay (unspecified)W 91% min
Assay (unspecified)C 5.9% min
Ethylenediaminetetraacetic acid, iron(III) monosodium salt is used to eliminate enzyme inhibition by traces of heavy metals and to inhibit enzymes that require divalent cations as cofactors.