Barium titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Barium titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Barium titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Barium titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Barium titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals
For vacuum deposition
Have Questions?
Catalog NumberQuantity
ALF088266.1425 g
Catalog number ALF088266.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS2316-64-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Infrared spectrumConforms
Melting point106°C to 111°C
Appearance (Form)Crystalline powder or flakes
GC>=97.5 %
Barium titanium oxide is used for vacuum deposition. It is also used in electronic ceramics, PTC thermistor, the configuration of the capacitors and as a raw material and intermediate in organic synthesis, pharmaceuticals and agrochemicals.